Learn More
Carbosynth LLC. MOG (Rat, Mouse, 35-55), 5mg, MW: (M.W. 2582.01), Molecular formula: C118H177N35O29S, CAS# [163913-87-9]
SDP
Supplier:  Carbosynth LLC. PMG3660PI5MG
Encompass_Preferred
Synonyms: H-Met-Glu-Val-Gly-Trp-Tyr-Arg-Ser-Pro-Phe-Ser-Arg-Val-Val-His-Leu-Tyr-Arg-Asn-Gly-Lys-OH Myelin Oligodendrocyte Protein (35-55), Application: Encephalitogenic Determinant, Storage Temperature: -20°C, References: M Ichikawa, T.G. Johns, J. Liu, and C.C. Bernard, J. Immunol. 157, 919-926 (1996), H.-C. von Büdingen, N. Tanuma, P. Villoslada, J.-C. Ouallet, S.L. Hauser, and C.P. Genain, J. Clin. Immunol., 21, 155 (2001).
Catalog No. 50-168-6664