Learn More
Medchemexpress LLC Mogroside I A1 | 88901-46-6 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN68541MG
Encompass_Preferred
Mogroside I A1 is a triterpenoid glycoside and a nonsugar sweetener. It is known for being sweeter than sucrose and exhibits antioxidant, antidiabetic, and anticancer activities.
  • Triterpenoid glycoside
  • Nonsugar sweetener, sweeter than sucrose
  • Exhibits antioxidant, antidiabetic, and anticancer activities
  • Store at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
Catalog No. 50-004-18278