Learn More
Medchemexpress LLC Mogroside I E1 | 88901-39-7 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN6853100MG
Encompass_Preferred
Mogroside I E1 is a triterpenoid glycoside that functions as a nonsugar sweetener, known to be sweeter than sucrose. This compound has demonstrated antioxidant, antidiabetic, and anticancer activities. It is intended for research use only. The product has a purity of 98.46% and appears as a white to off-white solid. Its molecular weight is 638.87 and its chemical formula is C36H62O9. It is sourced from *Cucurbitaceae Siraitia grosvenorii (Swingle) C. Jeffrey ex Lu et Z. Y. Zhang*.
Catalog No. 50-004-18367