Learn More
Medchemexpress LLC Mogroside I E1 | 88901-39-7 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN6853
Encompass_Preferred
Mogroside I E1 is a triterpenoid glycoside and a nonsugar sweetener. Mogrosides are sweeter than sucrose. Mogrosides exhibit antioxidant, antidiabetic, and anticancer activities.
  • Triterpenoid glycoside
  • Nonsugar sweetener
  • Sweeter than sucrose
  • Exhibits antioxidant activity
  • Exhibits antidiabetic activity
  • Exhibits anticancer activity
Catalog No. 50-004-20353