Learn More
Medchemexpress LLC HY-108272 5mg Medchemexpress, Mogroside II-A2 CAS:88901-45-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1082725MG
Encompass_Preferred
Medchemexpress, HY-108272 5mg Mogroside II-A2 CAS:88901-45-5 Mogroside II-A2, a triterpenoid glycoside isolated from the extracts of Luo Han Guo, is a nonsugar sweetener. Mogrosides are sweeter than sucrose. Mogrosides exhibit antioxidant, antidiabetic and anticancer activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0736