Learn More
Medchemexpress LLC Mogroside II-A2 | 88901-45-5 | MFCD21333504 | 99.9% | 801.01 g/mol | C42H72O14 | 1 MG

Supplier: Medchemexpress LLC HY1082721MG
Mogroside II-A2 is a triterpenoid glycoside isolated from Siraitia grosvenorii and supplied as a high-purity research standard for biochemical and preclinical studies. It is used as a nonsugar sweetener and is investigated for antioxidant, antidiabetic, and anticancer activities; supplied for formulation and assay work in small-mass packages.
- High purity (approximately 99.9%).
- Chemical formula C42H72O14; molecular weight 801.01 g/mol.
- Soluble in DMSO at about 100 mg/mL; sonication recommended.
- Storage: solid at 4°C protected from light; in solution store at -80°C up to 6 months.
- Suitable for in vitro assays and preclinical formulation studies.
- Available in small milligram packaging for research use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.