Learn More
Medchemexpress LLC Mogroside IIe | 88901-38-6 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN681410MM/1ML
Encompass_Preferred
Mogroside IIe is a nonsugar sweetener known for its sweetness, which surpasses that of sucrose. This compound is recognized for its antioxidant, antidiabetic, and anticancer properties.
  • Nonsugar sweetener
  • Sweeter than sucrose
  • Exhibits antioxidant activity
  • Exhibits antidiabetic activity
  • Exhibits anticancer activity
Catalog No. 50-004-19451