Learn More
Medchemexpress LLC Mogroside III-A1 | 88901-42-2 | 99.3% | 963.15 | C48H82O19 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1082711MG
Encompass_Preferred
Mogroside III-A1 is a cucurbitane-type triterpenoid glycoside isolated from Siraitia grosvenorii (monk fruit). It is used as a nonsugar sweetener and supplied as a purified research reagent for biochemical and pharmacological studies. The compound is investigated for antioxidant, antidiabetic, and anticancer activities and is provided with analytical characterization for research use.
Catalog No. 50-002-44073