Learn More
Medchemexpress LLC Mogroside IV | 89590-95-4 | 98.6% | 1125.29 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHYN6945
Encompass_Preferred
Mogroside IV is a triterpenoid glycoside isolated from Luo Han Guo extracts. It serves as a nonsugar sweetener, known to be sweeter than sucrose, and is derived from the plant Siraitia grosvenorii. This compound, along with other mogrosides, demonstrates various biological activities including antioxidant, antidiabetic, and anticancer properties.
  • Triterpenoid glycoside structure
  • Derived from Siraitia grosvenorii (Luo Han Guo)
  • Functions as a nonsugar sweetener
  • Sweeter than sucrose
  • Exhibits antioxidant properties
  • Demonstrates antidiabetic effects
  • Possesses anticancer activities
  • Appears as a white to off-white solid
Catalog No. 50-003-93341