Learn More
Medchemexpress LLC Mogroside IV-E | 88915-64-4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN245610MM/1ML
Encompass_Preferred
Mogroside IV-E is a triterpenoid glycoside and a nonsugar sweetener. Mogrosides are known to be sweeter than sucrose and exhibit antioxidant, antidiabetic, and anticancer activities.
  • Triterpenoid glycoside and nonsugar sweetener
  • Sweeter than sucrose
  • Exhibits antioxidant activity
  • Exhibits antidiabetic activity
  • Exhibits anticancer activity
Catalog No. 50-004-21081