Learn More
Medchemexpress LLC Mogroside IV-E | 88915-64-4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN24565MG
Encompass_Preferred
Mogroside IV-E is a triterpenoid glycoside that functions as a nonsugar sweetener. Mogrosides are known to be sweeter than sucrose.
  • Exhibits antioxidant, antidiabetic, and anticancer activities.
  • For research use only.
Catalog No. 50-004-21845