Learn More
Selleck Chemical LLC Mogroside V S3815-5mg
SDP
Supplier:  Selleck Chemical LLC S38155MG
Encompass_Preferred
Mogroside V is a natural cucurbitane glycoside which has a sweetening strength of 250 times that of sucrose and is derived from mature fruit of luo-han-guo (Siraitia grosvenorii monk fruit)
Catalog No. 50-311-0533