Learn More
Medchemexpress LLC Mogroside VI A | 2146088-13-1 | 5 MG

Supplier: Medchemexpress LLC HYN74385MG
Mogroside VI A is an isomer of Mogroside VI, isolated from Luo Han Guo. It exhibits sweetness properties and can be used for sweetener and/or taste modifier research. It has a molecular weight of 1449.58, a formula of C66H112O34, and appears as a white to off-white solid with a purity of 99.33%.
- Isomer of mogroside VI
- Isolated from luo han guo
- Possesses sweetness properties
- Utilized in research for sweeteners and/or taste modifiers
- Molecular weight of 1449.58
- Molecular formula: C66H112O34
- Appears as a white to off-white solid
- Purity of 99.33%
- Origin: Plants, cucurbitaceae, *Siraitia grosvenorii*
- Structure classification: Terpenoids, triterpenes
- Soluble in DMSO at 15 mg/mL (10.35 mM)
- Requires ultrasonic and warming for dissolution in DMSO
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.