Learn More
Medchemexpress LLC Mogroside VI B | 2149606-17-5 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN74391MG
Encompass_Preferred
Mogroside VI B is a cucurbitane glucoside separated from the crude extract of Siraitia grosvenorii. It shows an effect on activating PGC-1α transcription.
  • Separated from the crude extract of Siraitia grosvenorii
  • Activates PGC-1α transcription
  • Shows moderate effects on PGC-1α promoter activity
  • Exhibits PGC-1α promoter activity at 20 μM
Catalog No. 50-004-18047