Learn More
Medchemexpress LLC Mogroside VI B | 2149606-17-5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN74395MG
Encompass_Preferred
Mogroside VI B is a cucurbitane glucoside separated from the crude extract of Siraitia grosvenorii. It activates PGC-1α transcription and exhibits moderate PGC-1α promoter activity at 20 μM.
  • Cucurbitane glucoside
  • Isolated from Siraitia grosvenorii
  • Activates PGC-1α transcription
  • Moderate PGC-1α promoter activity at 20 μM
  • High purity (98.97%)
Catalog No. 50-004-19359