Learn More
Medchemexpress LLC Moguisteine (BBR-2173) | 119637-67-1 | 99.9% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYB0505
Encompass_Preferred
Moguisteine (BBR-2173) is an antitussive compound that does not exhibit significant affinity for opiate receptors. It is available as a solid, appearing white to off-white, and is intended for research use only. The product has a molecular weight of 339.41 and a chemical formula of C16H21NO5S.
  • Antitussive properties
  • Non-opiate: it has no significant affinity for opiate receptors, suggesting a different mechanism of action compared to opiate-based antitussives
  • High purity
  • Multiple formats: available in solid form and also as a 10 mM solution in DMSO for various research needs
  • Good solubility: for in vitro use, it has a solubility of 100 mg/mL in DMSO
  • Detailed documentation: data sheet, COA, SDS, and handling instructions are provided
  • Research use only: specifically designated for research applications
Catalog No. 50-004-06490