Learn More
Vee Gee Vee Gee Mohr Measuring Pipet, 10 mL, 0.1 graduation interval
Supplier:  Vee Gee 2010A10
Encompass_Preferred
The Mohr Measuring Pipet, delivers and drains out. It meets ASTM E-1293 Style 1, Class A Specifications and USP Standards for volumetric glassware.: 10.0 mL Tolerance: 0.03 mL Graduation Interval: 0.1 mL Approx. O.D. x Height: 10 x 370 mm Color Code: Orange
Catalog No. NC1937443