Learn More
Metrohm USA MOLECULAR SIEVE 6.2811.000 25O
Supplier: Metrohm USA 62811000
Molecular sieve. Bottle containing 250 g. Pore size: 0.3 nm. Without moisture indicator. For Rancimats and Karl Fischer instruments