Learn More
Sigma Aldrich Fine Chemicals Biosciences Molecular sieves 13X beads1KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 2086471KG
Encompass
Molecular sieves 13X showed preferential adsorption of CO over nitrogen or hydrogen at all pressures up to 250psia.
Catalog No. 50-289-9156