Learn More
Biolytic Lab Performance Inc Molecular Trap 20 gm for up to 4000 ml Bottle
SDP
Supplier:  Biolytic Lab Performance Inc 166101503
Encompass
Molecular Trap, 20 gram, for use in drying and maintaining dry solvents. The 20 gram size is good for up to 4000 ml Bottles. These molecular traps can be used to maintain anhydrous solvents on oligo synthesis, peptide synthesis and NMR.
Catalog No. NC1795837