Learn More
Selleck Chemical LLC Molibresib
SDP
Supplier:  Selleck Chemical LLC S718910MM/1ML
Encompass_Preferred
Molibresib (I-BET-762 GSK525762 GSK525762A) is an inhibitor for BET proteins with IC50 of 35 nM in a cell-free assay suppresses the production of proinflammatory proteins by macrophages and blocks acute inflammation highly selective over other bromodomain-containing proteins
Catalog No. 50-000-04090