Learn More
Medchemexpress LLC HY-12654 10mg Medchemexpress, Molidustat CAS:1154028-82-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1265410MG
Encompass_Preferred
Medchemexpress, HY-12654 10mg Molidustat CAS:1154028-82-6 Molidustat (BAY 85-3934) is a novel inhibitor of hypoxia-inducible factor prolyl hydroxylase (HIF-PH) with mean IC50 values of 480 nM for PHD1, 280 nM for PHD2, and 450 nM for PHD3. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8902
 Restricted Item