Learn More
Medchemexpress LLC Molindone hydrochloride | 15622-65-8 | 99.8% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB10175MG
Encompass_Preferred
Molindone hydrochloride (EN-1733A) is an orally active, brain-penetrant dopamine D2/D5 receptor antagonist. It demonstrates antipsychotic and antidepressant-like activities, suppresses spontaneous locomotion, and antagonizes apomorphine-induced emesis. This compound is suitable for research related to neurological diseases.
  • Orally active and brain-penetrant D2/D5 receptor antagonist
  • Exhibits antipsychotic and antidepressant-like activities
  • Suppresses spontaneous locomotion
  • Antagonizes apomorphine-induced emesis
  • Targeted to D2 and D5 receptors
  • Soluble in water (100 mg/mL; ultrasonic needed)
  • Soluble in DMSO (17.5 mg/mL; ultrasonic and warming needed)
Catalog No. 50-004-07532