Learn More
Medchemexpress LLC HY-135853 25mg , Molnupiravir CAS:2492423-29-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13585325MG
Encompass_Preferred
HY-135853 25mg Molnupiravir CAS: 2492423-29-5 Molnupiravir (EIDD-2801) is an orally bioavailable prodrug of the ribonucleoside analog EIDD-1931. Molnupiravir has broad spectrum antiviral activity against influenza virus and multiple coronaviruses, such as SARS-CoV-2, MERS-CoV, SARS-CoV. Molnupiravir has the potential for the research of COVID-19, and seasonal and pandemic influenza. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9140