Learn More
Medchemexpress LLC Molnupiravir-d7 | 00-00-0 | 99.9% | C13H12D7N3O7 | 10MG
SDP
Supplier:  Medchemexpress LLC HY135853S10MG
Encompass_Preferred
Molnupiravir-d7 is a deuterated isotopologue of the antiviral prodrug molnupiravir provided for research use only. It is intended as an isotopic internal standard, tracer, or analytical reference in pharmacokinetic, metabolic, and mass-spectrometry studies and is not for human or veterinary use.
  • Deuterated analog suitable as an isotopic internal standard.
  • High chemical purity (99.9%).
  • Useful for pharmacokinetic and metabolic tracer experiments.
  • Compatible with LC-MS/MS quantitation workflows.
  • Supplied in small-mass research pack sizes for analytical use.
Catalog No. 50-002-60043