Learn More
Strem, An Ascensus Company CAS# 34041-09-3. 100g. Molybdenum(IV) 2-ethylhexanoate (15% Mo). MFCD01631280
SDP
Supplier:  Strem, An Ascensus Company 421400100G
Encompass
CAS# 34041-09-3. 100g. Molybdenum(IV) 2-ethylhexanoate (15% Mo). MFCD01631280. Molecular Weight: 668.75. Molecular Formula: Mo[OOCCH(C2H5)C4H9]4. Color/form: reddish-brown liq. Strem# 42-1400. http://www.strem.com/catalog/v/42-1400/
Catalog No. 50-901-11712