Learn More
Sigma Aldrich Fine Chemicals Biosciences Molybdenum Blue spray reag
Supplier:  Sigma Aldrich Fine Chemicals Biosciences M1942100ML
Encompass
Molybdenum Blue reagent is suitable for use in the detection of phospholipids and related compounds.
Catalog No. 50-290-0946