Learn More
Strem, An Ascensus Company CAS# 18868-43-4. 50g. Molybdenum(IV) oxide, 99%. MFCD00014223
SDP
Supplier:  Strem, An Ascensus Company 93424650G
Encompass
CAS# 18868-43-4. 50g. Molybdenum(IV) oxide, 99%. MFCD00014223. Molecular Weight: 127.95. Molecular Formula: MoO2. Color/form: purple pwdr. Strem# 93-4246. http://www.strem.com/catalog/v/93-4246/
Catalog No. 50-901-15341