Learn More
Sigma Aldrich Fine Chemicals Biosciences Molybdenum Mo standard sol
Supplier:  Sigma Aldrich Fine Chemicals Biosciences NIST3134
Encompass
This Standard Reference Material (SRM) is intended for use as a primary calibration standard for the quantitative determination of molybdenum. A unit of SRM 3134 consists of five 10 mL sealed borosilicate glass ampoules of an acidified aqueous SOLUTIONon prepared gravimetrically to contain a known mass fraction of molybdenum. The SOLUTIONon contains hydrochloric acid at a volume fraction of approximately 10 % equivalent to an amount-of-substance concentration (molarity) of approximately 1.2 mol/L. For more information please refer ot the COA and SDS.SRM 3134 cert SRM 3134 SDS
Catalog No. 50-290-7246