Learn More
Strem, An Ascensus Company CAS# 17524-05-9. 25g. Molybdenum(VI) dioxide bis(acetylacetonate), min. 95%. MFCD00011506
SDP
Supplier:  Strem, An Ascensus Company 93421725G
Encompass
CAS# 17524-05-9. 25g. Molybdenum(VI) dioxide bis(acetylacetonate), min. 95%. MFCD00011506. Molecular Weight: 326.16. Molecular Formula: MoO2(CH3C(O)CHC(O)CH3)2. Color/form: yellow to orange pwdr. Strem# 93-4217. http://www.strem.com/catalog/v/93-4217/
Catalog No. 50-901-15335