Learn More
Medchemexpress LLC Momelotinib | 1056634-68-4 | MFCD16038899 | 98.9% | 414.46 | 50 MG

Supplier: Medchemexpress LLC HY10961200MG
Momelotinib (CYT387) is an ATP-competitive inhibitor of Janus Kinase (JAK) 1 and JAK2, with IC50 values of 11 nM and 18 nM, respectively. It exhibits significantly less activity against JAK3. This product complies with given specifications and is intended for research use only, as it has not been fully validated for medical applications.
- Selective JAK1/JAK2 inhibitor
- ATP-competitive mechanism
- Tested and complies with specifications
- For research use only
- Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
- Recommended storage for in solvent: -80°C for 2 years, -20°C for 1 year
- Appearance: Light yellow to yellow solid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.