Learn More
Medchemexpress LLC Mometasone | 105102-22-5 | 99.5% | 5 MG

Supplier: Medchemexpress LLC HYB06295MG
Mometasone is an inhaled glucocorticoid that can be used for mild asthma with a low sputum eosinophil level. It also shows potential for research in chronic hand eczema and rhinosinusitis.
- Inhaled glucocorticoid
- Can be used for mild asthma with a low sputum eosinophil level
- Potential for research in chronic hand eczema
- Potential for research in rhinosinusitis
- Appearance: White to off-white solid
- Purity: 99.46%
- Target: Glucocorticoid receptor
- Pathway: Immunology/inflammation; Vitamin D related/nuclear receptor
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.