Learn More
Medchemexpress LLC Momordicin IV | 894412-35-2 | 98.0% | 634.84 | C36H58O9 | 10 MG

Supplier: Medchemexpress LLC HYN1357910MG
Momordicin IV is a natural triterpenoid research compound provided as a purified reference material for biochemical and pharmacological studies. It appears as a white to off-white solid and is typically used in screening, structural characterization, and mechanism-of-action research.
- Natural triterpenoid compound.
- Intended for research use only; not for human or veterinary use.
- High purity: 98.0%.
- Chemical formula: C36H58O9.
- Molecular weight: 634.84 g/mol.
- CAS number: 894412-35-2.
- Available in small quantities, including a 10 MG pack suitable for initial studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.