Learn More
Cayman Chemical MonacetylphloroglucInolhyd 5g
SDP
Supplier:  Cayman Chemical 194515 G
Encompass_Preferred
A metabolite produced by fluorescent pseudomonads that is acetylated by MAPG acetyltransferase to form the broad-spectrum antibiotic, DAPG
Catalog No. 50-298-2687