Learn More
Cayman Chemical MonACtylcadavRInhydroch 500mg
SDP
Supplier:  Cayman Chemical 35275500 MG
Encompass_Preferred
A metabolite of cadaverine; plasma levels are increased in patients with schizophrenia
Catalog No. 50-291-3411