Learn More
New England Biolabs, Inc. Monarch Mag Beads M2 - 9 ml
SDP
Supplier:  New England Biolabs, Inc. T4105S
Encompass
Monarch Mag Beads M2 are designed to deliver high-performance nucleic acid purification and concentration. These magnetic beads are compatible with both manual and automated workflows, allowing easy integration and flexibility across a wide range of experimental setups. They are included as a component of Monarch DNA and RNA purification kits and are offered individually for convenience.
  • Monarch magnetic beads designed for nucleic acid purification
  • Silica-coated magnetic beads
  • Compatible with both manual and automated workflows, enabling flexible integration
  • Included as a component of Monarch DNA/RNA purification kits, offered individually for convenience
Catalog No. 50-003-32613