Learn More
Selleck Chemical LLC Monastrol
SDP
Supplier:  Selleck Chemical LLC S84391G
Encompass_Preferred
Monastrol (( )-Monastrol) is a cell-permeable small molecule inhibitor of kinesin-5(KIF11) with an IC50 of 14 M which is essential for maintaining separation of the half-spindles
Catalog No. 50-000-01688