Learn More
Medchemexpress LLC Monastrol | 329689-23-8 | MFCD11978057 | 292.35 g/mol | C14H16N2O3S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY101071A5MG
Encompass_Preferred
Monastrol is a cell-permeable, allosteric inhibitor of the mitotic kinesin Eg5 (KIF11) used in cell biology and cancer research to induce mitotic arrest and study spindle dynamics. It produces monopolar spindles and exhibits antiproliferative effects in multiple cell lines.
  • Selective inhibitor of the mitotic kinesin Eg5 (KIF11).
  • Cell-permeable small molecule suitable for in vitro and cellular assays.
  • Induces mitotic arrest characterized by monopolar spindles.
  • Reported IC50 ≈ 14 μM against Eg5 ATPase activity.
  • Supplied as a solid research reagent; common pack sizes include 1 mg, 5 mg, and 10 mg.
Catalog No. 50-002-12276