Learn More
Cayman Chemical MonbromobImn 100mg
SDP
Supplier:  Cayman Chemical 17097100 MG
Encompass_Preferred
A thiol-reactive fluorogenic probe; used to evaluate or quantify a variety of compounds containing reactive sulfur or thiol groups, including H2S, glutathione, proteins, and nucleotides
Catalog No. 50-296-5015