Learn More
Sigma Aldrich Fine Chemicals Biosciences Monensin sodium salt 90 955G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences M52735G
Encompass
Monensin is a polyether ionophoric antibiotic which is produced by Streptomyces cinnamonensis. It is used to treat bacterial fungal and parasitic infections. Monensin prevents the growth of colon cancer cells. It facilitates the transport of sodium and potassium ions between intracellular and extracellular spaces. Monensin prevents coccidiosisnbspin poultry production.
Catalog No. 50-290-2698