Learn More
Alpco Ltd Monkey Albumin ELISA
SDP
Supplier:  Alpco Ltd 41ALBMKE01
Encompass
The Albumin test kit is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring Albumin in Monkey serum, plasma, and urine. For Research Use Only. Not for Diagnostic Purposes.
Catalog No. NC2885138