Learn More
American Research Products Inc Monkey Brain deriVed neurotrop
Supplier:  American Research Products Inc CSBEL002655RH
Encompass
Monkey Brain-deriVed neurotrophic factor (BDNF) ELISA kit from CUSABIO Detection Range 0 78 ng/mL-50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26881