Learn More
Alpco Ltd Monkey Cystatin C ELISA
SDP
Supplier:  Alpco Ltd 41CYCMKE01
Encompass
Monkey Cystatin C ELISA is for the quantitative determination of Cystatin C in monkey plasma and serum. Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. NC2885194