Learn More
American Research Products Inc Monkey Immunoglobulin G IgG E
Supplier:  American Research Products Inc CSBE14412MK
Encompass
Monkey Immunoglobulin G IgG ELISA Kit from CUSABIO Detection Range 0 39 g/mL-100 g/mL Sensitivity 0 1 g/mL Sample serum plasma Method Competitive
Catalog No. 50-003-28494