Learn More
American Research Products Inc Monkey Insulin like Growth Fac
Supplier:  American Research Products Inc CSBE13401MK
Encompass
Monkey Insulin-like Growth Factor 1 (IGF-1) ELISA KIT from CUSABIO Detection Range 0 625 ng/ml-40 ng/ml Sensitivity 0 312 ng/ml Sample serum plasma Method Sandwich
Catalog No. 50-003-29393