Learn More
American Research Products Inc Monkey macrophage colony stimu
Supplier:  American Research Products Inc CSBEL006043MK
Encompass
Monkey macrophage colony-stimulating factor (M-CSF) ELISA kit from CUSABIO Detection Range 39 07 pg/mL-2500 pg/mL Sensitivity 9 77 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27753