Learn More
Abcam Monkey MMP3 ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB3153141X96TESTS
Encompass_Preferred
Monkey MMP3 ELISA Kit is a single-wash 90-min Simplestep used to quantify Monkey MMP3 with a sensitivity of 10.45 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Validated on a number of sample types including cerebrospinal fluid (CSF)
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-269-8354