Learn More
Cusabio Technology LLC Monkey N-terminal pro-brain natriuretic peptide NT-proBNP ELISA Kit, Sandwich ELISA, Detection range 0.78 ng/mL to 50 ng/mL, Sensitivity 0.195 ng/mL, Sample types: serum, plasma, tissue homogenates
Supplier:  Cusabio Technology LLC CSBE14327MK
Encompass_Preferred
Monkey N-terminal pro-brain natriuretic peptide NT-proBNP ELISA Kit, Sandwich ELISA, Detection range 0.78 ng/mL to 50 ng/mL, Sensitivity 0.195 ng/mL, Sample types: serum, plasma, tissue homogenates
Catalog No. NC3136545