Learn More
Genscript Corporation Monkeypox A29L protein His Ta
Supplier:  Genscript Corporation Z03752100
Encompass
The A29L protein from Monkeypox binds to cell surface heparin to promote fusion of viral membrane with host plasma membrane it is a potential target for developing diagnosis and vaccine
Catalog No. 50-000-57174