Learn More
Cayman Chemical Monmethyl AurIstatIn E 10mg
SDP
Supplier:  Cayman Chemical 1626710 MG
Encompass_Preferred
A synthetic analog of dolastatin 10 that inhibits tubulin polymerization and exhibits potent cytotoxicity; commonly conjugated with monoclonal antibodies directed at antigens specific to cancer cells for tumor-directed cytotoxicity
Catalog No. 50-297-2287